Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304485.1 | NP_001291414.1 | 4 | 447 | ||
NM_001304486.1 | NP_001291415.1 | 3 | 325 | |||
NM_001304487.1 | NP_001291416.1 | 3 | 359 | |||
NM_001304488.1 | NP_001291417.1 | 4 | 413 | |||
NM_001304489.1 | NP_001291418.1 | 4 | 413 | |||
NM_001304490.1 | NP_001291419.1 | 4 | 413 | |||
NM_001304491.1 | NP_001291420.1 | 3 | 325 | |||
NM_012460.3 | NP_036592.1 | 5 | 636 | |||
NR_130750.1 | 4 | 571 | ||||
NR_130751.1 | 3 | 348 | ||||
NR_130752.1 | 2 | 260 | ||||
GenBank mRNA | AF150100.1 | - | 4 | 327 | ||
AF152353.1 | - | 2 | 110 | |||
AK000666.1 | - | 4 | 345 | |||
AK312095.1 | - | 3 | 271 | |||
BC020213.1 | - | 5 | 536 | 4640518 | ||
BC054875.1 | - | 3 | 249 | 6663143 | ||
BF034731.1 | - | 4 | 291 | |||
BG249379.1 | - | 4 | 285 | |||
BM811095.1 | - | 4 | 483 | |||
HY041857.1 | - | 3 | 228 | |||
HY096567.1 | - | 3 | 277 | |||
W74224.1 | - | 4 | 316 |