Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PABPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002568.3 | NP_002559.2 | 3 | 998 | ||
XM_005250861.2 | XP_005250918.1 | 3 | 1909 | |||
GenBank mRNA | AK095194.1 | - | 3 | 833 | ||
AK130523.1 | - | 3 | 998 | |||
AK298990.1 | - | 4 | 714 | |||
AK303120.1 | - | 4 | 722 | |||
BC015958.2 | - | 4 | 893 | 3940309 | ||
BC023217.1 | - | 3 | 977 | |||
BC023520.2 | - | 3 | 977 | 4123269 | ||
BC041863.1 | - | 3 | 1622 | |||
Y00345.1 | - | 3 | 996 | |||
Z48501.1 | - | 3 | 419 |