Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014390.2 | NP_055205.2 | 14 | 1737 | ||
XM_017011987.1 | XP_016867476.1 | 14 | 1766 | |||
GenBank mRNA | AB209510.1 | - | 14 | 1675 | ||
AK096583.1 | - | 15 | 1662 | |||
AK304181.1 | - | 14 | 1629 | |||
AK309014.1 | - | 6 | 630 | |||
AK312313.1 | - | 14 | 1691 | |||
BC017180.2 | - | 14 | 1693 | 3345037 | ||
BE799345.1 | - | 1 | 73 | |||
BT009785.1 | - | 14 | 1511 | |||
U22055.1 | - | 14 | 1703 |