Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NMRK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289117.1 | NP_001276046.1 | 8 | 872 | ||
NM_170678.2 | NP_733778.1 | 8 | 857 | |||
NR_110316.1 | 5 | 438 | ||||
XM_006722725.3 | XP_006722788.1 | 6 | 711 | |||
XM_006722726.3 | XP_006722789.1 | 6 | 673 | |||
XM_017026597.1 | XP_016882086.1 | 6 | 653 | |||
GenBank mRNA | AF190819.1 | - | 6 | 508 | ||
AK001663.1 | - | 8 | 867 | |||
AK022514.1 | - | 7 | 699 | |||
AL365377.1 | - | 7 | 633 | |||
AY611481.1 | - | 7 | 567 | |||
BC093637.1 | - | 7 | 632 | 7939482 | ||
BC101575.1 | - | 7 | 632 | 8069081 | ||
BC143329.1 | - | 7 | 647 | 9051834 | ||
BM717225.1 | - | 5 | 425 |