Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PGAP2 SIRNA Assays ›
Gene Symbol:
PGAP2
Gene Name:
post-GPI attachment to proteins 2
Gene Aliases:
CWH43-N, FRAG1, HPMRS3, MRT17, MRT21
Chromosome Location:
Chr.11: 3797724 - 3826371 on Build GRCh38
UniGene ID:
Hs.133968
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s26141
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PGAP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145438.2
NP_001138910.1
4, 5
623
NM_001256235.1
NP_001243164.1
5, 6
564
NM_001256236.1
NP_001243165.1
5, 6
806
NM_001256237.1
NP_001243166.1
4, 5
623
NM_001256238.1
NP_001243167.1
4, 5
537
NM_001256239.1
NP_001243168.1
3, 4
553
NM_001256240.1
NP_001243169.1
3, 4
553
NM_001283038.1
NP_001269967.1
4, 5
623
NM_001283039.1
NP_001269968.1
4, 5
576
NM_014489.3
NP_055304.1
4, 5
736
NR_027016.2
2, 3
378
NR_027017.2
5, 6
686
NR_027018.2
5, 6
575
NR_045923.1
3, 4
448
NR_045927.1
3, 4
448
NR_045929.1
3, 4
506
NR_104270.1
3, 4
448
NR_104271.1
5, 6
708
NR_104272.1
2, 3
378
XM_006718181.3
XP_006718244.1
3, 4
1214
XM_006718183.3
XP_006718246.1
5, 6
631
XM_006718185.2
XP_006718248.1
4, 5
523
XM_006718186.2
XP_006718249.1
5, 6
683
XM_006718190.3
XP_006718253.1
4, 5
839
XM_006718191.3
XP_006718254.1
4, 5
839
XM_006718192.1
XP_006718255.1
6, 7
757
XM_006718193.2
XP_006718256.1
5, 6
564
XM_011519990.2
XP_011518292.2
5, 6
1090
XM_011519991.2
XP_011518293.2
5, 6
1093
XM_011519992.1
XP_011518294.1
6, 7
862
XM_011519993.2
XP_011518295.2
5, 6
1109
XM_011519994.2
XP_011518296.2
5, 6
1109
XM_011519995.2
XP_011518297.1
5, 6
1013
XM_011519996.1
XP_011518298.1
6, 7
814
XM_011519997.1
XP_011518299.1
6, 7
1155
XM_011519998.2
XP_011518300.1
6, 7
833
XM_011519999.1
XP_011518301.1
6, 7
792
XM_011520000.1
XP_011518302.1
5, 6
706
XM_011520001.1
XP_011518303.1
4, 5
498
XM_011520002.1
XP_011518304.1
4, 5
498
XM_011520003.2
XP_011518305.1
5, 6
1022
XM_011520004.2
XP_011518306.1
5, 6
697
XM_011520005.2
XP_011518307.1
4, 5
594
XM_011520006.2
XP_011518308.1
5, 6
978
XM_011520007.2
XP_011518309.1
6, 7
747
XM_017017556.1
XP_016873045.1
4, 5
498
XM_017017557.1
XP_016873046.1
4, 5
646
XM_017017558.1
XP_016873047.1
4, 5
820
XM_017017559.1
XP_016873048.1
5, 6
683
XM_017017560.1
XP_016873049.1
5, 6
683
GenBank mRNA
AF159615.1
-
4, 5
738
AK292181.1
-
5, 6
700
AK295202.1
-
5, 6
575
AK301991.1
-
5, 6
656
AK307359.1
-
3, 4
365
AL096753.1
-
5, 6
564
AY373030.1
-
2, 3
427
BC009930.2
-
4, 5
510
3506614
BG707484.1
-
3, 4
342
BI759334.1
-
3, 4
445
BI768071.1
-
3, 4
510
BI771208.1
-
2, 3
339
BI772237.1
-
4, 5
516
BI911526.1
-
5, 6
597
BX340489.2
-
3, 4
490
BX356273.2
-
4, 5
530
BX379511.2
-
4, 5
505
BX448155.2
-
4, 5
482
CB988453.1
-
2, 3
405
CD675009.1
-
1, 2
89
CD675113.1
-
3, 4
341
CX872317.1
-
2, 3
306
KU178608.1
-
2, 3
427
KU178609.1
-
3, 4
512
KU178610.1
-
3, 4
512
KU178611.1
-
2, 3
427
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001291358, NM_145583, AK159933, AK146442, BC004794, and others.
Panther Classification:
Molecular Function -
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
GPI anchor biosynthetic process
protein transport
signal transduction in response to DNA damage
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway in response to DNA damage
protein binding
protein transporter activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-9fjjm:80/10.239.165.64:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0