Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBMXL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162536.2 | NP_001156008.1 | 3 | 1471 | ||
NM_019610.5 | NP_062556.2 | 2 | 1371 | |||
GenBank mRNA | AL832554.1 | - | 2 | 1224 | ||
AL832785.1 | - | 1 | 315 | |||
BC012942.1 | - | 2 | 1030 | 3875565 |
Gene Symbol: RBMX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164803.1 | NP_001158275.1 | 5 | 640 | ||
NM_002139.3 | NP_002130.2 | 7 | 965 | |||
NR_028476.1 | 6 | 793 | ||||
NR_028477.1 | 7 | 1000 | ||||
GenBank mRNA | AK091520.1 | - | 6 | 739 | ||
AK096015.1 | - | 2 | 871 | |||
AK304573.1 | - | 7 | 945 | |||
AK304868.1 | - | 5 | 585 | |||
AK309911.1 | - | 7 | 920 | |||
AY464692.1 | - | 6 | 755 | |||
BC006550.2 | - | 7 | 901 | 3010123 | ||
BC007435.2 | - | 7 | 901 | 3010123 | ||
BX647131.1 | - | 6 | 736 | |||
Z23064.1 | - | 6 | 766 |