Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286756.1 | NP_001273685.1 | 27 | 2195 | ||
NM_013367.2 | NP_037499.2 | 27 | 2192 | |||
XM_005248159.2 | XP_005248216.1 | 14 | 1089 | |||
XM_011513838.1 | XP_011512140.1 | 27 | 2072 | |||
XM_017008138.1 | XP_016863627.1 | 27 | 2121 | |||
GenBank mRNA | AF191338.1 | - | 26 | 2063 | ||
AK001881.1 | - | 5 | 1850 | |||
AK023597.1 | - | 24 | 1806 | |||
AK292336.1 | - | 27 | 2112 | |||
AL353932.1 | - | 7 | 2310 | |||
BC059383.1 | - | 27 | 2165 | 30349524 |