Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021044.3 | NP_066382.1 | 2 | 745 | ||
XM_017019380.1 | XP_016874869.1 | 2 | 676 | |||
XM_017019381.1 | XP_016874870.1 | 2 | 393 | |||
GenBank mRNA | AK313360.1 | - | 2 | 715 | ||
BC033507.1 | - | 2 | 745 | 5169286 | ||
HQ607677.1 | - | 2 | 742 | |||
HQ607678.1 | - | 2 | 749 | |||
U59748.1 | - | 2 | 188 |