Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DACT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079520.1 | NP_001072988.1 | 4 | 2476 | ||
NM_016651.5 | NP_057735.2 | 4 | 2587 | |||
NR_046093.1 | 4 | 2085 | ||||
NR_046095.1 | 4 | 2393 | ||||
XM_006720167.3 | XP_006720230.1 | 4 | 2524 | |||
GenBank mRNA | AF251079.2 | - | 1 | 1692 | ||
AK299879.1 | - | 4 | 2393 | |||
AK316164.1 | - | 4 | 2183 | |||
AK316169.1 | - | 2 | 1799 | |||
AK316237.1 | - | 4 | 2085 | |||
AK316423.1 | - | 3 | 1952 | |||
BX161433.1 | - | 4 | 2040 |