Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290007.1 | NP_001276936.1 | 2 | 213 | ||
NM_001290008.1 | NP_001276937.1 | 2 | 222 | |||
NM_001290009.1 | NP_001276938.1 | 2 | 222 | |||
NM_016293.3 | NP_057377.3 | 2 | 222 | |||
XM_005268957.3 | XP_005269014.2 | 2 | 198 | |||
XM_005268958.3 | XP_005269015.1 | 2 | 222 | |||
GenBank mRNA | AB032698.1 | - | 2 | 164 | ||
AF146531.1 | - | 2 | 137 | |||
AK000783.1 | - | 2 | 178 | |||
AK093638.1 | - | 2 | 149 | |||
AK225911.1 | - | 2 | 178 | |||
AK300211.1 | - | 2 | 201 | |||
AK314304.1 | - | 2 | 198 | |||
BC047686.1 | - | 2 | 188 | 5722815 | ||
CB996605.1 | - | 2 | 240 | |||
DB035315.1 | - | 2 | 222 |