Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPHLN1 SIRNA Assays ›
Gene Symbol:
PPHLN1
Gene Name:
periphilin 1
Gene Aliases:
CR, HSPC206, HSPC232
Chromosome Location:
Chr.12: 42326126 - 42448621 on Build GRCh38
UniGene ID:
Hs.444157
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s28290
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPHLN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001143787.1
NP_001137259.1
8
806
NM_001143788.1
NP_001137260.1
8
914
NM_001143789.1
NP_001137261.1
7
749
NM_016488.6
NP_057572.5
9
971
NM_201438.1
NP_958846.1
9
849
NM_201439.1
NP_958847.1
9
971
NM_201440.1
NP_958848.1
9
849
NM_201515.1
NP_958923.1
10
1014
XM_005268962.3
XP_005269019.1
7
768
XM_011538459.2
XP_011536761.1
10
992
XM_011538465.2
XP_011536767.1
8
892
XM_011538467.2
XP_011536769.1
9
831
XM_011538470.2
XP_011536772.1
8
792
XM_011538471.2
XP_011536773.1
7
768
XM_017019434.1
XP_016874923.1
10
992
XM_017019435.1
XP_016874924.1
10
991
XM_017019436.1
XP_016874925.1
10
992
XM_017019437.1
XP_016874926.1
9
949
XM_017019438.1
XP_016874927.1
9
949
XM_017019439.1
XP_016874928.1
10
992
XM_017019440.1
XP_016874929.1
9
935
XM_017019441.1
XP_016874930.1
8
892
XM_017019442.1
XP_016874931.1
8
892
XM_017019443.1
XP_016874932.1
9
935
XM_017019444.1
XP_016874933.1
9
831
XM_017019445.1
XP_016874934.1
8
825
XM_017019446.1
XP_016874935.1
8
824
XM_017019447.1
XP_016874936.1
8
792
XM_017019448.1
XP_016874937.1
8
792
XM_017019449.1
XP_016874938.1
7
768
XM_017019450.1
XP_016874939.1
7
768
XM_017019451.1
XP_016874940.1
8
792
XM_017019452.1
XP_016874941.1
9
935
XM_017019453.1
XP_016874942.1
8
792
GenBank mRNA
AF151040.1
-
9
810
AF151066.1
-
9
824
AK000186.1
-
3
364
AK055690.1
-
9
959
AK129910.1
-
10
1014
AK299951.1
-
7
731
AK303612.1
-
8
806
AK303680.1
-
6
687
AK308022.1
-
8
888
AY039238.1
-
9
953
AY157850.1
-
10
1010
BC025306.1
-
8
773
BC039832.1
-
9
816
6051282
BC040449.1
-
9
816
DC424443.1
-
7
734
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
keratinization
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-zvggx:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0