Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202429.1 | NP_001189358.1 | 8, 9 | 1985 | ||
NM_016150.4 | NP_057234.2 | 6, 7 | 1960 | |||
XM_005267758.3 | XP_005267815.1 | 9, 10 | 2045 | |||
XM_011536834.2 | XP_011535136.1 | 7, 8 | 1895 | |||
XM_011536835.2 | XP_011535137.1 | 6, 7 | 1534 | |||
XM_017021369.1 | XP_016876858.1 | 7, 8 | 1944 | |||
GenBank mRNA | AB056723.1 | - | 6, 7 | 1960 | ||
AB488462.1 | - | 8, 9 | 1985 | |||
AF159164.1 | - | 6, 7 | 1342 | |||
AJ251238.1 | - | 6, 7 | 1614 | |||
AK002049.1 | - | 6, 7 | 1776 | |||
AK057078.1 | - | 9, 10 | 1933 | |||
AK093271.1 | - | 1, 2 | 1386 | |||
AK303686.1 | - | 8, 9 | 1804 | |||
AK315628.1 | - | 6, 7 | 1586 | |||
AL137735.1 | - | 6, 7 | 1381 | |||
BC032354.1 | - | 6, 7 | 1593 | 5218997 |