Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPGEF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164386.1 | NP_001157858.1 | 21 | 3395 | ||
NM_001164387.1 | NP_001157859.1 | 22 | 3410 | |||
NM_001164388.1 | NP_001157860.1 | 21 | 3395 | |||
NM_001164389.1 | NP_001157861.1 | 21 | 3395 | |||
NM_016340.5 | NP_057424.3 | 21 | 3395 | |||
GenBank mRNA | AF394782.1 | - | 22 | 3386 | ||
AF478468.1 | - | 21 | 3189 | |||
AF478469.1 | - | 21 | 3189 | |||
AF478567.1 | - | 21 | 3395 | |||
AK302640.1 | - | 21 | 3372 | |||
BC133703.1 | - | 21 | 3318 | 40124898 | ||
BC140820.1 | - | 21 | 3270 | 9021688 | ||
BC144627.1 | - | 21 | 3270 | 9053158 |