Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FXYD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022006.1 | NP_071289.1 | 2 | 134 | ||
GenBank mRNA | AI929519.1 | - | 2 | 134 | ||
AK057825.1 | - | 2 | 109 | |||
AK296405.1 | - | 2 | 128 | |||
BC018619.1 | - | 2 | 118 | 3626060 |