Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024885.3 | NP_079161.3 | 1 | 214 | ||
XM_006724664.1 | XP_006724727.1 | 1 | 213 | |||
GenBank mRNA | AK308261.1 | - | 1 | 215 | ||
BC043391.1 | - | 1 | 200 | 6154641 | ||
DB455298.1 | - | 1 | 214 |