Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RC3H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100588.1 | NP_001094058.1 | 11 | 2054 | ||
NM_018835.2 | NP_061323.2 | 11 | 2054 | |||
GenBank mRNA | AF255303.1 | - | 10 | 2414 | ||
AK000308.1 | - | 9 | 1488 | |||
AK027042.1 | - | 2 | 291 | |||
AK226010.1 | - | 9 | 1488 | |||
AL833177.1 | - | 12 | 1911 | |||
AL834431.1 | - | 5 | 805 |