Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019069.3 | NP_061942.2 | 1 | 748 | ||
GenBank mRNA | AK002149.1 | - | 1 | 476 | ||
AK315186.1 | - | 1 | 366 | |||
BC025762.1 | - | 1 | 748 | |||
BC043494.1 | - | 1 | 337 | 6056052 | ||
CR457279.1 | - | 1 | 241 |