Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018066.3 | NP_060536.3 | 2, 3 | 859 | ||
GenBank mRNA | AK001211.1 | - | 2, 3 | 778 | ||
AK022535.1 | - | 2, 3 | 764 | |||
AY358864.1 | - | 2, 3 | 768 | |||
AY364261.1 | - | 2, 3 | 577 | |||
BC007815.2 | - | 2, 3 | 701 | 4299794 | ||
BC008634.1 | - | 2, 3 | 725 | 3463813 | ||
BC020272.1 | - | 2, 3 | 744 |