Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS13C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018088.2 | NP_001018098.1 | 4 | 311 | ||
NM_017684.4 | NP_060154.3 | 4 | 311 | |||
NM_018080.3 | NP_060550.2 | 4 | 311 | |||
NM_020821.2 | NP_065872.1 | 4 | 311 | |||
XM_011521713.2 | XP_011520015.1 | 4 | 272 | |||
XM_011521714.2 | XP_011520016.1 | 4 | 272 | |||
GenBank mRNA | AB037842.1 | - | 4 | 294 | ||
AJ608770.1 | - | 4 | 294 | |||
AJ608771.1 | - | 4 | 294 | |||
AJ626860.1 | - | 4 | 294 | |||
AJ626861.1 | - | 4 | 294 | |||
AK307518.1 | - | 4 | 236 |