Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017957.2 | NP_060427.2 | 2 | 662 | ||
GenBank mRNA | AF324241.1 | - | 1 | 227 | ||
AK000785.1 | - | 2 | 611 | |||
AK291658.1 | - | 2 | 611 | |||
AK292977.1 | - | 2 | 611 | |||
AK295730.1 | - | 3 | 388 | |||
AK296346.1 | - | 3 | 529 | |||
BC001038.2 | - | 2 | 591 | |||
BC051365.1 | - | 2 | 591 | |||
BC077722.1 | - | 2 | 591 | |||
BC110602.2 | - | 2 | 275 | |||
BC110605.2 | - | 1 | 357 | 40027860 |