Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018079.4 | NP_060549.4 | 20 | 2765 | ||
XM_011532946.1 | XP_011531248.1 | 20 | 2718 | |||
XM_017004418.1 | XP_016859907.1 | 12 | 2319 | |||
GenBank mRNA | AK001241.1 | - | 14 | 1662 | ||
AK055896.1 | - | 5 | 1585 | |||
AK056536.1 | - | 20 | 2735 | |||
AK301122.1 | - | 12 | 1464 | |||
AK316222.1 | - | 12 | 1445 | |||
BC032538.1 | - | 14 | 1740 | 5528417 |