Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267571.1 | NP_001254500.1 | 11 | 2581 | ||
NM_001267572.1 | NP_001254501.1 | 5 | 1360 | |||
NM_018421.3 | NP_060891.3 | 11 | 2581 | |||
NR_051978.1 | 10 | 2439 | ||||
XM_011518843.1 | XP_011517145.1 | 6 | 2463 | |||
XM_017014899.1 | XP_016870388.1 | 11 | 2011 | |||
GenBank mRNA | AB449882.1 | - | 11 | 2401 | ||
AF318370.1 | - | 11 | 2440 | |||
AK026105.1 | - | 10 | 1929 | |||
AK124772.1 | - | 10 | 3130 | |||
AK298575.1 | - | 12 | 2231 | |||
AL353935.1 | - | 6 | 2642 | |||
AY026527.1 | - | 11 | 2493 | |||
BC028918.1 | - | 5 | 1360 | 4931447 | ||
BC071978.1 | - | 7 | 1468 | 5443205 |