Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267571.1 | NP_001254500.1 | 5 | 1077 | ||
NM_018421.3 | NP_060891.3 | 5 | 1077 | |||
NR_051978.1 | 4 | 935 | ||||
XM_011518844.2 | XP_011517146.1 | 5 | 1077 | |||
XM_017014899.1 | XP_016870388.1 | 3 | 379 | |||
GenBank mRNA | AB209721.1 | - | 5 | 877 | ||
AB449882.1 | - | 5 | 897 | |||
AF318370.1 | - | 5 | 936 | |||
AK026105.1 | - | 4 | 425 | |||
AK124772.1 | - | 4 | 1626 | |||
AK131290.1 | - | 5 | 955 | |||
AK298575.1 | - | 6 | 727 | |||
AY026527.1 | - | 5 | 989 |