Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018509.3 | NP_060979.2 | 4, 5 | 666 | ||
GenBank mRNA | AF119857.1 | - | 1, 2 | 24 | ||
AK025328.1 | - | 4, 5 | 666 | |||
AK316593.1 | - | 2, 3 | 168 | |||
BC017168.1 | - | 4, 5 | 624 | 3907072 | ||
BC052279.1 | - | 4, 5 | 584 | 4127952 |