Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018154.2 | NP_060624.1 | 4 | 628 | ||
GenBank mRNA | AB104486.1 | - | 4 | 456 | ||
AF279307.1 | - | 4 | 549 | |||
AK001288.1 | - | 4 | 587 | |||
AK001466.1 | - | 4 | 619 | |||
AK223080.1 | - | 4 | 582 | |||
AK302134.1 | - | 4 | 538 | |||
BC007726.2 | - | 4 | 552 | 4303318 | ||
BC010014.2 | - | 4 | 562 | 4304756 | ||
BC036521.1 | - | 4 | 592 | 5270216 | ||
CR457235.1 | - | 4 | 456 |