Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005751.2 | NP_001005751.1 | 27 | 2944 | ||
XM_005269806.3 | XP_005269863.2 | 26 | 2820 | |||
XM_005269807.3 | XP_005269864.2 | 26 | 2805 | |||
XM_005269810.3 | XP_005269867.2 | 9 | 1037 | |||
XM_006717831.3 | XP_006717894.2 | 26 | 2739 | |||
XM_017016227.1 | XP_016871716.1 | 26 | 2769 | |||
XM_017016230.1 | XP_016871719.1 | 26 | 2757 | |||
XM_017016233.1 | XP_016871722.1 | 24 | 2467 | |||
XM_017016234.1 | XP_016871723.1 | 26 | 2827 | |||
XM_017016235.1 | XP_016871724.1 | 18 | 1843 | |||
GenBank mRNA | AK001686.1 | - | 24 | 2596 | ||
AK304116.1 | - | 27 | 2880 | |||
BC082258.1 | - | 27 | 2871 | 6278433 | ||
CN314884.1 | - | 4 | 269 |
Gene Symbol: FAM21C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011539566.1 | XP_011537868.1 | 27 | 2911 | ||
XM_011539567.1 | XP_011537869.1 | 26 | 2839 | |||
XM_011539568.1 | XP_011537870.1 | 26 | 2824 | |||
XM_011539569.1 | XP_011537871.1 | 26 | 2758 | |||
XM_011539570.2 | XP_011537872.1 | 26 | 2800 | |||
XM_011539573.2 | XP_011537875.1 | 9 | 1037 | |||
XM_017016014.1 | XP_016871503.1 | 26 | 2788 | |||
XM_017016017.1 | XP_016871506.1 | 25 | 2752 | |||
XM_017016019.1 | XP_016871508.1 | 25 | 2686 | |||
XM_017016021.1 | XP_016871510.1 | 27 | 2839 | |||
XM_017016024.1 | XP_016871513.1 | 15 | 1653 | |||
XM_017016025.1 | XP_016871514.1 | 16 | 1724 | |||
GenBank mRNA | AK001545.1 | - | 9 | 1105 | ||
AK293929.1 | - | 22 | 2395 |