Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other HDAC8 SIRNA Assays ›
Gene Symbol:
HDAC8
Gene Name:
histone deacetylase 8
Gene Aliases:
CDA07, CDLS5, HD8, HDACL1, MRXS6, RPD3, WTS
Chromosome Location:
Chr.X: 72329516 - 72573103 on Build GRCh38
UniGene ID:
Hs.310536
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s31698
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: HDAC8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001166418.1
NP_001159890.1
1
392
NM_001166419.1
NP_001159891.1
1
392
NM_001166420.1
NP_001159892.1
1
392
NM_001166422.1
NP_001159894.1
1
392
NM_001166448.1
NP_001159920.1
1
392
NM_018486.2
NP_060956.1
1
392
NR_051952.1
1
392
XM_011530986.2
XP_011529288.2
1
387
XM_011530987.2
XP_011529289.2
1
387
XM_017029640.1
XP_016885129.1
1
387
XM_017029641.1
XP_016885130.1
1
387
XM_017029642.1
XP_016885131.1
1
385
XM_017029643.1
XP_016885132.1
1
387
XM_017029644.1
XP_016885133.1
1
387
XM_017029645.1
XP_016885134.1
1
387
XM_017029646.1
XP_016885135.1
1
67
XM_017029647.1
XP_016885136.1
1
387
GenBank mRNA
AF212246.1
-
1
108
AF230097.1
-
1
105
AF245664.1
-
1
59
AJ277724.1
-
1
92
AK074326.1
-
1
337
AK294903.1
-
1
106
AK296641.1
-
1
342
AK298767.1
-
1
132
AK300895.1
-
1
89
AY429539.1
-
1
67
BC050433.1
-
1
83
5761745
BP229332.1
-
1
368
BQ189619.1
-
1
142
BQ632790.1
-
1
104
DC352390.1
-
1
393
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00218503_m1
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
chromatin assembly or disassembly
transcription, DNA-templated
sister chromatid cohesion
chromatin modification
negative regulation of protein ubiquitination
regulation of protein stability
regulation of telomere maintenance
histone H3 deacetylation
regulation of cohesin loading
histone deacetylase activity
protein binding
transcription factor binding
Hsp70 protein binding
NAD-dependent histone deacetylase activity (H3-K14 specific)
metal ion binding
Hsp90 protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-c74kb:80/10.239.167.51:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0