Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206647.1 | NP_001193576.1 | 3 | 352 | ||
NM_020642.3 | NP_065693.2 | 3 | 352 | |||
NR_045417.1 | 2 | 534 | ||||
XM_017018011.1 | XP_016873500.1 | 3 | 372 | |||
GenBank mRNA | AF493783.1 | - | 3 | 352 | ||
AF493784.1 | - | 3 | 352 | |||
AF512007.1 | - | 2 | 276 | |||
AK294549.1 | - | 2 | 513 | |||
BC030996.2 | - | 2 | 282 | 4609949 | ||
BP355293.1 | - | 2 | 534 |