Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142648.1 | NP_001136120.1 | 7 | 629 | ||
NM_020150.4 | NP_064535.1 | 6 | 559 | |||
GenBank mRNA | AF086166.1 | - | 2 | 185 | ||
AF087897.1 | - | 6 | 526 | |||
AF217959.1 | - | 6 | 526 | |||
AF261717.1 | - | 6 | 526 | |||
AF274026.1 | - | 6 | 525 | |||
AF318066.1 | - | 5 | 426 | |||
AK298591.1 | - | 5 | 392 | |||
AL136724.1 | - | 6 | 542 | |||
AY008268.1 | - | 6 | 550 | |||
BC003658.2 | - | 6 | 524 | 3546972 | ||
BG327663.1 | - | 7 | 597 | |||
CR533492.1 | - | 5 | 426 |