Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTUD7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020205.3 | NP_064590.2 | 8 | 1228 | ||
XM_011509782.1 | XP_011508084.1 | 8 | 1225 | |||
XM_011509783.2 | XP_011508085.1 | 8 | 1455 | |||
XM_011509784.2 | XP_011508086.1 | 8 | 1019 | |||
XM_011509785.2 | XP_011508087.1 | 8 | 4652 | |||
XM_011509786.1 | XP_011508088.1 | 8 | 1091 | |||
XM_011509787.1 | XP_011508089.1 | 8 | 834 | |||
XM_011509788.1 | XP_011508090.1 | 7 | 943 | |||
XM_017001850.1 | XP_016857339.1 | 8 | 1091 | |||
XM_017001851.1 | XP_016857340.1 | 7 | 1077 | |||
GenBank mRNA | AJ293573.1 | - | 8 | 1071 | ||
AK299790.1 | - | 8 | 1214 | |||
BC020622.1 | - | 8 | 1014 | |||
BC072681.1 | - | 8 | 1165 | 6170513 |