Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278596.1 | NP_001265525.1 | 25, 26 | 2686 | ||
NM_001278597.1 | NP_001265526.1 | 25, 26 | 2704 | |||
XM_006718988.3 | XP_006719051.1 | 23, 24 | 2961 | |||
XM_011520975.2 | XP_011519277.1 | 23, 24 | 5247 | |||
XM_011520978.2 | XP_011519280.1 | 22, 23 | 2559 | |||
XM_017019672.1 | XP_016875161.1 | 23, 24 | 2961 | |||
GenBank mRNA | AJ272204.1 | - | 25, 26 | 2704 | ||
AK024010.1 | - | 8, 9 | 968 | |||
BM710588.1 | - | 1, 2 | 118 | |||
FJ384095.1 | - | 25, 26 | 2660 |