Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGLYRP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020393.3 | NP_065126.2 | 7 | 1099 | ||
XM_011509789.2 | XP_011508091.1 | 7 | 1111 | |||
XM_011509790.1 | XP_011508092.1 | 7 | 1099 | |||
XM_011509791.2 | XP_011508093.1 | 7 | 1090 | |||
XM_011509792.1 | XP_011508094.1 | 7 | 1099 | |||
XM_011509793.2 | XP_011508095.1 | 4 | 787 | |||
GenBank mRNA | AF242518.1 | - | 7 | 812 | ||
AK292203.1 | - | 7 | 1102 | |||
AY035377.1 | - | 7 | 804 | |||
BC107157.1 | - | 6 | 786 | 40032748 | ||
BC107158.1 | - | 6 | 786 | 40032750 | ||
BC142636.1 | - | 7 | 1074 | 40147772 |