Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYRM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020466.4 | NP_065199.1 | 2 | 142 | ||
NR_028493.1 | 2 | 284 | ||||
NR_028494.1 | 2 | 284 | ||||
NR_028495.1 | 2 | 152 | ||||
GenBank mRNA | AK024783.1 | - | 2 | 140 | ||
AK094994.1 | - | 2 | 193 | |||
AK311151.1 | - | 2 | 284 | |||
AK311180.1 | - | 2 | 201 | |||
AK311275.1 | - | 2 | 137 | |||
AK311948.1 | - | 2 | 127 | |||
AL122090.1 | - | 2 | 125 | |||
AV710669.1 | - | 2 | 142 | |||
BC009782.1 | - | 2 | 136 | 4092710 | ||
BC104668.1 | - | 2 | 125 | 6717725 | ||
BP280644.1 | - | 2 | 142 | |||
DA160441.1 | - | 2 | 284 |