Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020715.2 | NP_065766.1 | 20 | 2950 | ||
XM_011537009.1 | XP_011535311.1 | 20 | 3126 | |||
XM_011537015.2 | XP_011535317.1 | 13 | 3181 | |||
XM_017021499.1 | XP_016876988.1 | 19 | 3107 | |||
XM_017021500.1 | XP_016876989.1 | 20 | 3126 | |||
XM_017021501.1 | XP_016876990.1 | 19 | 3023 | |||
XM_017021502.1 | XP_016876991.1 | 13 | 2903 | |||
XM_017021503.1 | XP_016876992.1 | 13 | 2263 | |||
XM_017021504.1 | XP_016876993.1 | 15 | 2405 | |||
GenBank mRNA | AB033026.1 | - | 20 | 2926 | ||
AK293950.1 | - | 1 | 28 | |||
AK295896.1 | - | 13 | 1650 | |||
BC146787.1 | - | 20 | 2926 | 9007246 | ||
BX648464.1 | - | 11 | 2062 |