Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other GRAMD1B SIRNA Assays ›
Gene Symbol:
GRAMD1B
Gene Name:
GRAM domain containing 1B
Gene Aliases:
-
Chromosome Location:
Chr.11: 123358299 - 123627771 on Build GRCh38
UniGene ID:
Hs.144725
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s33114
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: GRAMD1B
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001286563.1
NP_001273492.1
20
2583
NM_001286564.1
NP_001273493.1
19
2292
NM_020716.2
NP_065767.1
19
2562
XM_005271619.4
XP_005271676.1
18
2550
XM_005271621.4
XP_005271678.1
19
6937
XM_005271623.4
XP_005271680.1
18
2315
XM_005271624.4
XP_005271681.2
18
2230
XM_006718892.3
XP_006718955.1
19
2467
XM_011542926.2
XP_011541228.1
19
2464
XM_011542929.2
XP_011541231.1
18
2440
XM_011542930.2
XP_011541232.1
18
3081
XM_011542931.2
XP_011541233.1
18
2995
XM_011542933.1
XP_011541235.1
17
2704
XM_017018034.1
XP_016873523.1
20
2535
XM_017018035.1
XP_016873524.1
20
2532
XM_017018036.1
XP_016873525.1
19
3002
XM_017018037.1
XP_016873526.1
20
3301
XM_017018038.1
XP_016873527.1
21
2547
XM_017018039.1
XP_016873528.1
19
2593
XM_017018040.1
XP_016873529.1
18
2389
XM_017018041.1
XP_016873530.1
20
2605
XM_017018042.1
XP_016873531.1
19
2593
XM_017018043.1
XP_016873532.1
18
2226
XM_017018044.1
XP_016873533.1
18
2550
XM_017018045.1
XP_016873534.1
19
6934
XM_017018046.1
XP_016873535.1
19
3039
XM_017018047.1
XP_016873536.1
20
3048
XM_017018048.1
XP_016873537.1
19
3004
XM_017018049.1
XP_016873538.1
19
3033
XM_017018050.1
XP_016873539.1
18
2309
XM_017018051.1
XP_016873540.1
17
1925
GenBank mRNA
AB033027.1
-
19
2225
AI815988.1
-
1
38
AK127457.1
-
20
2583
AK297606.1
-
18
2280
AY358924.1
-
8
1803
BC105741.1
-
19
2376
40026783
BC105931.1
-
19
2376
40026781
BC105932.1
-
19
2376
40026782
BC107480.1
-
19
2376
40026784
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-gk48b:80/10.239.167.222:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0