Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHROOM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020859.3 | NP_065910.3 | 5 | 2172 | ||
XM_005263163.4 | XP_005263220.1 | 4 | 6626 | |||
XM_005263164.4 | XP_005263221.1 | 4 | 1184 | |||
XM_005263165.4 | XP_005263222.1 | 4 | 2610 | |||
XM_006714282.3 | XP_006714345.1 | 5 | 1327 | |||
XM_011532157.2 | XP_011530459.2 | 4 | 2014 | |||
XM_011532158.2 | XP_011530460.1 | 6 | 1973 | |||
XM_011532159.1 | XP_011530461.1 | 5 | 2162 | |||
XM_011532160.1 | XP_011530462.1 | 3 | 1174 | |||
XM_011532161.2 | XP_011530463.1 | 4 | 1352 | |||
XM_017008488.1 | XP_016863977.1 | 5 | 1330 | |||
XM_017008489.1 | XP_016863978.1 | 5 | 1231 | |||
GenBank mRNA | AB055660.1 | - | 5 | 2173 | ||
AK128677.1 | - | 1 | 674 | |||
AK304103.1 | - | 4 | 968 |