Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C12orf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021640.3 | NP_067653.3 | 4 | 622 | ||
GenBank mRNA | AF111805.1 | - | 2 | 214 | ||
AF289485.1 | - | 4 | 590 | |||
BC028904.1 | - | 3 | 526 | 4454943 | ||
BC051871.1 | - | 4 | 581 | 5170816 |