Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EDA2R SIRNA Assays ›
Gene Symbol:
EDA2R
Gene Name:
ectodysplasin A2 receptor
Gene Aliases:
EDA-A2R, EDAA2R, TNFRSF27, XEDAR
Chromosome Location:
Chr.X: 66594384 - 66639303 on Build GRCh38
UniGene ID:
Hs.302017
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s34058
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EDA2R
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001199687.2
NP_001186616.1
6
647
NM_001242310.1
NP_001229239.1
6
612
NM_001324199.1
NP_001311128.1
4
293
NM_001324201.1
NP_001311130.1
5
567
NM_001324202.1
NP_001311131.1
4
291
NM_001324204.1
NP_001311133.1
4
390
NM_001324205.1
NP_001311134.1
4
384
NM_001324206.1
NP_001311135.1
6
655
NM_021783.4
NP_068555.1
6
649
NR_136726.1
6
556
NR_136727.1
6
558
XM_011531000.1
XP_011529302.1
7
662
XM_011531001.1
XP_011529303.1
7
710
XM_011531002.1
XP_011529304.1
7
662
XM_011531003.2
XP_011529305.1
5
462
XM_011531004.1
XP_011529306.1
7
656
XM_011531005.2
XP_011529307.1
4
296
XM_011531007.2
XP_011529309.1
7
605
XM_017029703.1
XP_016885192.1
5
559
XM_017029704.1
XP_016885193.1
6
599
XM_017029705.1
XP_016885194.1
5
437
XM_017029707.1
XP_016885196.1
5
346
XM_017029708.1
XP_016885197.1
4
283
GenBank mRNA
AF298812.1
-
5
539
AK296264.1
-
6
508
AK299018.1
-
4
249
AK314881.1
-
6
615
AY152724.1
-
6
602
AY358735.1
-
5
548
AY358736.1
-
5
542
BC034919.1
-
6
596
5434545
DC368837.1
-
4
249
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
multicellular organism development
epidermis development
tissue development
ectodermal cell differentiation
tumor necrosis factor-mediated signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
positive regulation of JNK cascade
positive regulation of NF-kappaB transcription factor activity
intrinsic apoptotic signaling pathway by p53 class mediator
receptor activity
tumor necrosis factor-activated receptor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-kqhf4:80/10.239.162.79:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0