Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174159.1 | NP_001167630.1 | 7 | 1073 | ||
NM_001174160.1 | NP_001167631.1 | 6 | 838 | |||
NM_022071.3 | NP_071354.2 | 7 | 1223 | |||
XM_011544610.1 | XP_011542912.1 | 6 | 1269 | |||
XM_017013729.1 | XP_016869218.1 | 6 | 991 | |||
GenBank mRNA | AK024620.1 | - | 5 | 492 | ||
AK024799.1 | - | 7 | 1101 | |||
AK225052.1 | - | 5 | 492 | |||
AK293301.1 | - | 6 | 714 | |||
BC014525.2 | - | 4 | 419 | |||
BC067117.1 | - | 4 | 433 | |||
BC082982.1 | - | 7 | 1073 | 6575224 |