Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022143.4 | NP_071426.1 | 2 | 525 | ||
XM_011516461.2 | XP_011514763.1 | 3 | 831 | |||
XM_017012505.1 | XP_016867994.1 | 3 | 721 | |||
GenBank mRNA | AF196976.4 | - | 2 | 528 | ||
AJ297858.1 | - | 2 | 528 | |||
AK172751.1 | - | 2 | 529 | |||
AK314047.1 | - | 2 | 601 | |||
AY358307.1 | - | 1 | 444 | |||
BC111561.1 | - | 2 | 519 | 40068142 | ||
BC111745.1 | - | 2 | 519 | 40068146 |