Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040284.2 | NP_001035374.2 | 5 | 862 | ||
NM_001040285.2 | NP_001035375.2 | 5 | 862 | |||
XM_005256092.4 | XP_005256149.1 | 4 | 1045 | |||
XM_005256093.4 | XP_005256150.2 | 5 | 859 | |||
XM_011523275.2 | XP_011521577.1 | 5 | 858 | |||
XM_011523276.2 | XP_011521578.1 | 5 | 860 | |||
XM_011523278.2 | XP_011521580.1 | 4 | 333 | |||
XM_017023555.1 | XP_016879044.1 | 5 | 918 | |||
GenBank mRNA | AK001141.1 | - | 1 | 62 | ||
AK097589.1 | - | 5 | 677 | |||
AK223410.1 | - | 3 | 132 | |||
AK300918.1 | - | 5 | 862 | |||
AL833922.1 | - | 2 | 105 | |||
DN994465.1 | - | 4 | 480 | |||
FR872509.1 | - | 4 | 626 |