Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282640.1 | NP_001269569.1 | 4 | 546 | ||
NM_001282643.1 | NP_001269572.1 | 4 | 546 | |||
NM_022486.4 | NP_071931.2 | 4 | 546 | |||
XM_011518915.1 | XP_011517217.1 | 4 | 769 | |||
XM_011518916.2 | XP_011517218.1 | 4 | 769 | |||
XM_011518917.1 | XP_011517219.1 | 4 | 769 | |||
XM_011518918.1 | XP_011517220.1 | 4 | 769 | |||
XM_011518919.1 | XP_011517221.1 | 3 | 613 | |||
XM_017015037.1 | XP_016870526.1 | 4 | 769 | |||
XM_017015038.1 | XP_016870527.1 | 4 | 769 | |||
XM_017015039.1 | XP_016870528.1 | 4 | 769 | |||
XM_017015040.1 | XP_016870529.1 | 4 | 769 | |||
GenBank mRNA | AK056704.1 | - | 4 | 434 | ||
AK292868.1 | - | 4 | 546 | |||
AY358746.1 | - | 4 | 525 | |||
BC060770.1 | - | 4 | 516 | 30348765 | ||
BX362255.2 | - | 4 | 534 | |||
DB221801.1 | - | 4 | 547 |