Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NADK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198993.1 | NP_001185922.1 | 4, 5 | 553 | ||
NM_001198995.1 | NP_001185924.1 | 2, 3 | 415 | |||
NM_023018.4 | NP_075394.3 | 4, 5 | 622 | |||
XM_005244778.2 | XP_005244835.1 | 4, 5 | 683 | |||
XM_006710837.2 | XP_006710900.1 | 5, 6 | 798 | |||
XM_006710838.2 | XP_006710901.1 | 4, 5 | 531 | |||
XM_006710839.1 | XP_006710902.1 | 7, 8 | 1110 | |||
XM_017002143.1 | XP_016857632.1 | 4, 5 | 683 | |||
XM_017002144.1 | XP_016857633.1 | 2, 3 | 427 | |||
GenBank mRNA | AK023114.1 | - | 4, 5 | 622 | ||
AK290161.1 | - | 4, 5 | 604 | |||
AK291599.1 | - | 4, 5 | 546 | |||
AK291728.1 | - | 4, 5 | 522 | |||
AK296722.1 | - | 2, 3 | 415 | |||
AY090771.1 | - | 3, 4 | 400 | |||
BC001709.1 | - | 4, 5 | 485 | 3542247 |