Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CAPRIN2 SIRNA Assays ›
Gene Symbol:
CAPRIN2
Gene Name:
caprin family member 2
Gene Aliases:
C1QDC1, EEG-1, EEG1, RNG140
Chromosome Location:
Chr.12: 30709552 - 30754994 on Build GRCh38
UniGene ID:
Hs.234355
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s35296
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CAPRIN2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002259.2
NP_001002259.1
17
3519
NM_001206856.2
NP_001193785.1
16
3351
NM_001319842.1
NP_001306771.1
16
3433
NM_001319843.1
NP_001306772.1
17
3516
NM_001319844.1
NP_001306773.1
17
2669
NM_001319845.1
NP_001306774.1
18
2774
NM_001319846.1
NP_001306775.1
18
2764
NM_023925.4
NP_076414.2
16
3369
NM_032156.4
NP_115532.3
17
3516
XM_006719138.3
XP_006719201.1
17
3579
XM_006719139.3
XP_006719202.1
16
3537
XM_006719140.3
XP_006719203.1
16
3534
XM_006719142.3
XP_006719205.1
16
3432
XM_006719144.3
XP_006719207.1
15
3390
XM_006719145.3
XP_006719208.1
17
3642
XM_006719146.3
XP_006719209.1
17
3642
XM_006719147.3
XP_006719210.1
17
3579
XM_017019851.1
XP_016875340.1
16
3474
XM_017019852.1
XP_016875341.1
18
2734
XM_017019853.1
XP_016875342.1
18
2804
XM_017019854.1
XP_016875343.1
15
3387
XM_017019855.1
XP_016875344.1
18
2701
XM_017019856.1
XP_016875345.1
17
2659
XM_017019857.1
XP_016875346.1
17
2629
XM_017019858.1
XP_016875347.1
17
2656
XM_017019859.1
XP_016875348.1
16
2485
XM_017019860.1
XP_016875349.1
18
3686
XM_017019861.1
XP_016875350.1
16
2509
XM_017019862.1
XP_016875351.1
18
3683
XM_017019863.1
XP_016875352.1
18
3686
XM_017019864.1
XP_016875353.1
18
3686
XM_017019865.1
XP_016875354.1
16
3537
XM_017019866.1
XP_016875355.1
18
2737
XM_017019867.1
XP_016875356.1
18
2764
XM_017019868.1
XP_016875357.1
18
2734
XM_017019869.1
XP_016875358.1
18
2761
XM_017019870.1
XP_016875359.1
18
2674
XM_017019871.1
XP_016875360.1
18
2745
XM_017019872.1
XP_016875361.1
17
2596
XM_017019873.1
XP_016875362.1
16
2512
XM_017019874.1
XP_016875363.1
17
2651
XM_017019875.1
XP_016875364.1
16
2449
XM_017019876.1
XP_016875365.1
15
2202
XM_017019877.1
XP_016875366.1
14
2160
XM_017019878.1
XP_016875367.1
14
2157
XM_017019879.1
XP_016875368.1
16
2486
XM_017019880.1
XP_016875369.1
13
1950
XM_017019881.1
XP_016875370.1
17
2651
XM_017019882.1
XP_016875371.1
17
2591
XM_017019883.1
XP_016875372.1
14
2160
GenBank mRNA
AB058776.1
-
2962
AF525416.1
-
9
1030
AK024247.1
-
4
2486
AK026222.1
-
3
335
AL834212.1
-
15
2047
AY074490.1
-
16
3369
AY074491.1
-
16
3433
BC007520.1
-
3
240
BC066295.1
-
3
227
BC111007.1
-
3
279
BC117672.1
-
17
3313
40068103
BC117673.1
-
16
3148
40068104
BX537569.1
-
14
5531
BX538080.1
-
4
411
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of translation
negative regulation of cell growth
positive regulation of protein binding
positive regulation of peptidyl-serine phosphorylation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of dendrite morphogenesis
positive regulation of dendritic spine morphogenesis
positive regulation of canonical Wnt signaling pathway
RNA binding
receptor binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-f2ldd:80/10.239.165.14:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0