Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TMUB2 SIRNA Assays ›
Gene Symbol:
TMUB2
Gene Name:
transmembrane and ubiquitin like domain containing 2
Gene Aliases:
FP2653
Chromosome Location:
Chr.17: 44186956 - 44191731 on Build GRCh38
UniGene ID:
Hs.181391
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s35561
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TMUB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001076674.1
NP_001070142.1
4
855
NM_024107.2
NP_077012.2
3
787
NM_177441.2
NP_803190.2
3
1288
XM_011525198.2
XP_011523500.1
3
1191
XM_011525200.2
XP_011523502.1
5
883
XM_011525201.2
XP_011523503.1
4
863
XM_011525203.2
XP_011523505.1
5
981
XM_011525204.2
XP_011523506.1
4
920
XM_011525205.2
XP_011523507.1
4
1381
XM_011525206.2
XP_011523508.1
5
1067
XM_011525207.2
XP_011523509.1
4
999
XM_011525208.2
XP_011523510.1
3
892
XM_011525209.2
XP_011523511.1
3
868
XM_011525210.2
XP_011523512.1
6
957
XM_011525211.2
XP_011523513.1
4
714
XM_017025047.1
XP_016880536.1
4
1294
XM_017025048.1
XP_016880537.1
4
964
XM_017025049.1
XP_016880538.1
3
1278
XM_017025050.1
XP_016880539.1
3
892
XM_017025051.1
XP_016880540.1
3
810
XM_017025052.1
XP_016880541.1
5
1183
XM_017025053.1
XP_016880542.1
6
870
XM_017025054.1
XP_016880543.1
4
1167
XM_017025055.1
XP_016880544.1
4
754
XM_017025056.1
XP_016880545.1
4
592
GenBank mRNA
AF289564.1
-
3
470
AF370373.1
-
3
1265
AK294198.1
-
6
953
AL533563.3
-
4
850
AL833899.1
-
3
1288
AY007092.1
-
3
710
AY358780.1
-
3
761
BC001792.1
-
3
512
BC004306.1
-
3
434
BC063489.1
-
3
761
5239969
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-kt7sl:80/10.239.164.98:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0