Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083924.1 | NP_001077393.1 | 3 | 1110 | ||
NM_023938.5 | NP_076427.2 | 4 | 1296 | |||
XM_005273259.1 | XP_005273316.1 | 2 | 864 | |||
XM_006711530.1 | XP_006711593.1 | 4 | 1173 | |||
XM_011509973.2 | XP_011508275.1 | 3 | 966 | |||
GenBank mRNA | AK093826.1 | - | 4 | 1264 | ||
AL832940.1 | - | 1 | 370 | |||
AY352640.1 | - | 4 | 1287 | |||
BC000765.2 | - | 4 | 1192 | 2822914 | ||
BC018959.2 | - | 2 | 953 |