Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024334.2 | NP_077310.1 | 11 | 1148 | ||
XM_017007176.1 | XP_016862665.1 | 12 | 1270 | |||
GenBank mRNA | AK027466.1 | - | 5 | 378 | ||
AK027757.1 | - | 11 | 1074 | |||
AK027827.1 | - | 6 | 431 | |||
AK027877.1 | - | 11 | 1012 | |||
AK074073.1 | - | 9 | 3579 | |||
AK075010.1 | - | 11 | 999 | |||
AK225589.1 | - | 11 | 1071 | |||
AL136916.1 | - | 11 | 1042 | |||
AY358625.1 | - | 11 | 911 | |||
BC003125.1 | - | 11 | 1043 | 3503636 | ||
BC008054.2 | - | 9 | 638 | |||
BC011719.2 | - | 11 | 1043 | 4079766 | ||
KU178769.1 | - | 10 | 894 | |||
KU178770.1 | - | 9 | 1468 |