Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP13A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024524.3 | NP_078800.3 | 9 | 1202 | ||
XM_005269357.3 | XP_005269414.1 | 9 | 1223 | |||
XM_011513120.2 | XP_011511422.1 | 10 | 1324 | |||
XM_011513121.1 | XP_011511423.1 | 9 | 1202 | |||
XM_011513122.1 | XP_011511424.1 | 9 | 896 | |||
XM_011513123.2 | XP_011511425.1 | 10 | 1329 | |||
XM_011513124.2 | XP_011511426.1 | 10 | 1329 | |||
GenBank mRNA | BP313821.1 | - | 3 | 249 | ||
DR005713.1 | - | 2 | 83 |