Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IGFLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024660.2 | NP_078936.1 | 3 | 277 | ||
GenBank mRNA | AK026226.1 | - | 3 | 278 | ||
AX775837.1 | - | 3 | 280 | |||
BC126481.1 | - | 3 | 238 | 8992197 | ||
BC126483.1 | - | 3 | 238 | 8992199 | ||
BC144205.1 | - | 3 | 238 |