siRNA Details
Gene Symbol: VCAN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126336.2 | NP_001119808.1 | 6 | 1233 | ||
NM_001164097.1 | NP_001157569.1 | 6 | 1233 | |||
NM_001164098.1 | NP_001157570.1 | 6 | 1233 | |||
NM_004385.4 | NP_004376.2 | 6 | 1233 | |||
GenBank mRNA | AB209491.1 | - | 6 | 1116 | ||
AK299770.1 | - | 2 | 151 | |||
AK307642.1 | - | 6 | 1134 | |||
BC050524.1 | - | 6 | 1116 | |||
D32039.1 | - | 6 | 982 | |||
U16306.1 | - | 6 | 1143 | |||
U26555.1 | - | 6 | 1143 | |||
X15998.1 | - | 6 | 1143 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
